missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human p300 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3789-SP
This item is not returnable.
View return policy
Description
KAT3B/p300 Polyclonal specifically detects KAT3B/p300 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Chromatin Immunoprecipitation (ChIP), Immunocytochemistry.
Specifications
| KAT3B/p300 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| E1A binding protein p300, E1A-associated protein p300, E1A-binding protein, 300kD, EC 2.3.1, EC 2.3.1.48, EP300, histone acetyltransferase p300, KAT3B, P300, p300 HAT, RSTS2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2033 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| ChIP Assay, Immunocytochemistry, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 50 ug/mL, Immunohistochemistry 5-25 ug/mL, Chromatin Immunoprecipitation (ChIP) 5 ug/5 x 10^6 cells, Immunocytochemistry 1-15 ug/mL | |
| Q09472 | |
| EP300 | |
| E. coli-derived recombinant human p300 His2283-His2414 Accession # Q09472 | |
| 25 μg | |
| Primary | |
| Detects human p300 in Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction