missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human P2Y11/P2RY11 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB9305S-100UG
This item is not returnable.
View return policy
Description
P2Y11/P2RY11 Monoclonal specifically detects P2Y11/P2RY11 in Human samples. It is validated for Flow Cytometry.
Specifications
| P2Y11/P2RY11 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| P2RY11, P2Y11P2Y purinoceptor 11, purinergic receptor P2Y, G-protein coupled, 11, purinergic receptor P2Y11 | |
| NS0 mouse myeloma cell line transfected with human P2Y11/P2RY11 Accession # Q96G91 | |
| RUO | |
| 5032 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 505214 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q96G91 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Stains human P2Y11/P2RY11 transfectants but not irrelevant transfectants in flow cytometry. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction