missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human P2X7/P2RX7 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB9790-SP
This item is not returnable.
View return policy
Description
P2X7/P2RX7 Monoclonal specifically detects P2X7/P2RX7 in Human samples. It is validated for Immunocytochemistry.
Specifications
| P2X7/P2RX7 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| ATP receptor, MGC20089, P2RX7, P2X purinoceptor 7, P2X7P2X7 receptor, P2X7R, P2Z receptor, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 7, purinergic receptor P2X7 variant A | |
| E. coli-derived recombinant human P2X7/P2RX7 Asp356-Tyr595 Accession # Q99572 | |
| 25 μg | |
| 5027 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Immunocytochemistry | |
| 545230 | |
| Immunocytochemistry 5-25 ug/mL | |
| Q99572 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human P2X7/P2RX7 in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction