missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech™ Human P21 ELISA Kit
Shop All Proteintech Products
Click to view available options
Target Species:
Human
Unit Size:
96 tests
Description
- CDKN1A (p21, CIP1, WAF1) a cyclin-dependent kinase inhibitor is necessary for proper control of cell cycle and premature senescence
- p21 is a cyclin-dependent kinase (CDK) inhibitor that suppresses proliferation by inhibiting CDK2 and CDK1 activity at G1/s and G2/M transitions
- Mediator of p53 to induce cell cycle arrest in G1
- Can interact with proliferating cell nuclear antigen, plays regulatory role in S phase DNA replication and DNA damage repair
- Overexpression decreases expression levels of cell cycle progression genes and upregulation senescence-inducing genes
- p21 is a cyclin-dependent kinase (CDK) inhibitor that suppresses proliferation by inhibiting CDK2 and CDK1 activity at G1/s and G2/M transitions
Process:
- Antibody specific for P21 pre-coated onto microwells
- P21 protein in samples captured by the coated antibody after incubation
- After extensive washing, another antibody specific for P21 is added to detect captured P21 protein
- For signal development, horseradish peroxidase (HRP)-conjugated antibody is added, followed by Tetramethyl-benzidine (TMB) reagent
- Solution containing sulfuric acid used to stop color development and color intensity
- Proportional to quantity of bound protein, measurable at 450 nm with correction wavelength set at 630 nm
Specifications
Specifications
| Accession Number | NP_000380 |
| Assay Range | 0.25 to 16 ng/mL |
| Assay Sensitivity | 0.1 ng/mL |
| Certifications/Compliance | ISO 13485 |
| Detection Method | Sandwich ELISA, Double Antibody |
| Immunoassay Kit Format | Coated ELISA Kit |
| Sample Type | Plasma, Serum |
| Substrate | DAB |
| Target Species | Human |
| Gene ID (Entrez) | 1026 |
| Show More |
Safety and Handling
Product Identifier
- 96TEST Human P21 ELISA Kit
- Warning
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P234-Keep only in original container.
- P264-Wash skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P330-Rinse mouth.
- P362-Take off contaminated clothing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction