missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human p16-INK4a Recombinant Protein, PeproTech®

Product Code. 30088482
Click to view available options
Quantity:
10 x 100 μg
100 μg
20 μg
5 μg
5 x 100 μg
Unit Size:
100µg
1mg
20µg
500µg
5µg
Pack of 10
Pack of 5
This item is not returnable. View return policy

Product Code. 30088482

Brand: Gibco™ 11002500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

110-02-1MG will be provided as 10 x 100 μg (110-02-100UG). 110-02-500UG will be provided as 5 x 100 μg (110-02-100UG). Recombinant Human p16-INK4a is a 16.5 kDa protein containing 156 amino acid residues. Kim, W.Y. and Sharpless, N.E. (2006)Cell 127(2):265-75. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This gene generates several transcript variants which differ in their first exons. At least three alternatively spliced variants encoding distinct proteins have been reported, two of which encode structurally related isoforms known to function as inhibitors of CDK4 kinase. The remaining transcript includes an alternate first exon located 20 Kb upstream of the remainder of the gene; this transcript contains an alternate open reading frame (ARF) that specifies a protein which is structurally unrelated to the products of the other variants. This ARF product functions as a stabilizer of the tumor suppressor protein p53 as it can interact with, and sequester, the E3 ubiquitin-protein ligase MDM2, a protein responsible for the degradation of p53. In spite of the structural and functional differences, the CDK inhibitor isoforms and the ARF product encoded by this gene, through the regulatory roles of CDK4 and p53 in cell cycle G1 progression, share a common functionality in cell cycle G1 control. This gene is frequently mutated or deleted in a wide variety of tumors, and is known to be an important tumor suppressor gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P42771, Q8N726
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 1029
Molecular Weight (g/mol) 16.5 kDa
Name Human p16-INK4a
Quantity 5 x 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias Alternative reading frame; Arf; ARF-INK4a; CDK4 inhibitor p16 INK4; CDK4 inhibitor p16-INK4; CDK4I; CDKN2; Cdkn2a; cdkn2a {ECO:0000312; cell cycle inhibitor; cell cycle negative regulator beta; cell cycle regulator; CMM2; Cyclin dependent kinase 4 inhibitor A; cyclin dependent kinase inhibitor 2 A; Cyclin dependent kinase inhibitor 2 A (p16, inhibits CDK4); cyclin-dependent kinase 4 inhibitor A; Cyclin-dependent kinase inhibitor 2 A; cyclin-dependent kinase inhibitor 2 A (melanoma, p16, inhibits CDK4); cyclin-dependent kinase inhibitor 2 A (p16, inhibits CDK4); cyclin-dependent kinase inhibitor 2 A p16Ink4a; cyclin-dependent kinase inhibitor 2 A p19Arf; cyclin-dependent kinase inhibitor 2 A, isoform 1; cyclin-dependent kinase inhibitor 2 A, isoform 2; cyclin-dependent kinase inhibitor 2 A, isoform 3; cyclin-dependent kinase inhibitor 2 A, isoforms 1/2; cyclin-dependent kinase inhibitor protein; EMBL:AAL76336.1, ECO:0000312; EMBL:AAL76338.1, ECO:0000312; HGNC:1787; inhibits CDK4; INK4; INK4A; Ink4a/Arf; INK4a-ARF; isoforms 1/2/3; Melanoma p16 inhibits CDK4; mitochondrial smARF; MLM; MTS1; MTS-1; Multiple tumor suppressor 1; P14; P14 P16; P14ARF; p16; p16(INK4a); p16Cdkn2a; P16INK4; p16-INK4; p16INK4a; p16-INK4a; P19; P19 TP16; P19ARF; Pctr1; RGD:2323}; TP16; Tumor suppressor ARF
Common Name p16INK4a
Gene Symbol CDKN2A
Conjugate Unconjugated
Recombinant Recombinant
Sequence MEPAAGSSME PSADWLATAA ARGRVEEVRA LLEAGALPNA PNSYGRRPIQ VMMMGSARVA ELLLLHGAEP NCADPATLTR PVHDAAREGF LDTLVVLHRA GARLDVRDAW GRLPVDLAEE LGHRDVARYL RAAAGGTRGS NHARIDAAEG PSDIPD
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.