missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human p15INK4b/CDKN2B Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6798
This item is not returnable.
View return policy
Description
p15INK4b/CDKN2B Monoclonal specifically detects p15INK4b/CDKN2B in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| p15INK4b/CDKN2B | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 3-25 ug/mL | |
| P42772 | |
| CDKN2B | |
| E. coli-derived recombinant human p15INK4b/CDKN2B Arg2-Asp138 Accession # P42772 | |
| 100 μg | |
| Primary | |
| Detects human p15INK4b/CDKN2B in direct ELISAs and Western blots. In Western blots, approximately 50% cross-reactivity with recombinant human (rh) p16INK4a and no cross-reactivity with rhp18INK4c or rhp19INK4d is observed. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 651308 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CDK4I, CDKN2B, cyclin-dependent kinase inhibitor 2B (p15, inhibits CDK4), cyclin-dependent kinases 4 and 6 binding protein, INK4BCDK4B inhibitor, MTS2, MTS-2, MTS2CDK inhibitory protein, Multiple tumor suppressor 2, p14_CDK inhibitor, p14_INK4B, p14-INK4b, p15 CDK inhibitor, p15_INK4B, P15cyclin-dependent kinase 4 inhibitor B, p15INK4b, p15-INK4b, TP15 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1030 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction