missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human P-Cadherin Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF861-SP
This item is not returnable.
View return policy
Description
P-Cadherin Polyclonal specifically detects P-Cadherin in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, CyTOF-ready.
Specifications
| P-Cadherin | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CAD3, cadherin 3, P-cadherin (placental), cadherin 3, type 1, P-cadherin (placental), cadherin-3, CDH3, CDHP, CDHPcalcium-dependent adhesion protein, placental, HJMD, PCAD, PCadherin, PCADP-cadherin, Placental cadherin | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1001 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| CAA45177 | |
| CDH3 | |
| Mouse myeloma cell line NS0-derived recombinant human P-Cadherin Asp108-Gly654 Accession # CAA45177 | |
| 25 μg | |
| Primary | |
| Detects human P-Cadherin in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) Cadherin-11, rhCadherin-12, rhCadherin-13, rhE-Cadherin and less than 1% cross-reactivity with rhCadherin-8, rhCadherin-17, rhN-Cadherin, rhK-Cadherin and recombinant mouse P-Cadherin is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction