missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human P-Cadherin Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB861-100
This item is not returnable.
View return policy
Description
P-Cadherin Monoclonal specifically detects P-Cadherin in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunocytochemistry, CyTOF-ready, Knockout Validated, ELISA Capture (Matched Antibody Pair).
Specifications
| P-Cadherin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CAD3, cadherin 3, P-cadherin (placental), cadherin 3, type 1, P-cadherin (placental), cadherin-3, CDH3, CDHP, CDHPcalcium-dependent adhesion protein, placental, HJMD, PCAD, PCadherin, PCADP-cadherin, Placental cadherin | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1001 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF, KnockDown, ELISA | |
| 104805 | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready, Knockout Validated 0.25 ug/10^6 cells, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| CAA45177 | |
| CDH3 | |
| Mouse myeloma cell line NS0-derived recombinant human P-Cadherin Asp108-Gly654 Accession # CAA45177 | |
| 100 μg | |
| Primary | |
| Detects human P-Cadherin in direct ELISAs and Western blots. In Western blots, does not cross-react with recombinant human (rh) Cadherin-8, recombinant mouse P-Cadherin, or rhVE-Cadherin. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction