missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human P-Cadherin Alexa Fluor 647-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB861RR-100UG
This item is not returnable.
View return policy
Description
P-Cadherin Monoclonal specifically detects P-Cadherin in Human samples. It is validated for Flow Cytometry.
Specifications
| P-Cadherin | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/ml in a saline solution containing BSA and Sodium Azide. | |
| CAD3, cadherin 3, P-cadherin (placental), cadherin 3, type 1, P-cadherin (placental), cadherin-3, CDH3, CDHP, CDHPcalcium-dependent adhesion protein, placental, HJMD, PCAD, PCadherin, PCADP-cadherin, Placental cadherin | |
| Mouse myeloma cell line NS0-derived recombinant human P-Cadherin Asp108-Gly654 Accession # CAA45177 | |
| 100 μg | |
| 1001 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 104805R | |
| Flow Cytometry 0.25-1 μg/10^6 cells | |
| CAA45177 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human P-Cadherin in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction