missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The Human OX40/TNFRSF4 ELISA Kit (Colorimetric) measures human OX40/TNFRSF4 in cell culture supernatant, serum.
Specifications
Specifications
| Assay Range | 78 - 5,000pg/mL |
| Assay Sensitivity | 1.8pg/mL |
| Product Type | ELISA Kit |
| Sample Type | Cell Culture Supernatant, Serum |
| Specificity | The assay detects both natural and recombinant human OX40/TNFRSF4. The cross reactivity of circulating factors of the immune system was evaluated by spiking these proteins at physiologically relevant concentrations into a human OX40/TNFRSF4 positive serum. There was no crossreactivity detected. |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 7293 |
| Gene Symbol | TNFRSF4 |
| Kit Contents | Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human sCD134/OX40, Biotin-Conjugate anti-human sCD134/OX40 monoclonal antibody, Streptavidin-HRP, Human sCD134/OX40 Standard lyophilized, 10ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?