missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human OX40 Ligand (soluble) Recombinant Protein, PeproTech®

Product Code. 17852363
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
2µg
500µg
This item is not returnable. View return policy

Product Code. 17852363

Brand: Gibco™ 31028250UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant OX40L is a glycosylated 133 amino acid protein corresponding to the extracellular TNF homologous domain of the full length transmembrane protein. It migrates with an apparent molecular mass of 15.5-25.0 kDa on SDS-PAGE. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The tumor necrosis factor superfamily member TNFSF4 is a type II membrane bound, non-covalently linked homotrimeric protein. It is expressed on antigen presenting cells, such as dendritic cells and activated B-cells, and also on other cells such as vascular endothelial cells, mast cells, and natural killer cells. TNFSF4 signals specifically through the TNFRSF4 receptor, is expressed predominantly on CD4+T cells but also on certain activated CD8+T cells. TNFRSF4/TNFSF4 functions as a costimulatory signal, which is required for a productive interaction between antigen presenting cells and their target T-cells. It enhances cell proliferation and survival, and increases expression of RANTES, IL-2, IL-3, and IFNgamma. TNFRSF4/TNFSF4 signaling plays an important role in immunotolerance.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P23510
For Use With (Application) Control, ELISA Standard, Binding Assay, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 7292
Molecular Weight (g/mol) 15.4 kDa
Name Human OX40 Ligand (soluble)
Purification Method Purified
Quantity 250 μg
Source Insect cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias Ath1; Ath-1; atherosclerosis 1; CD134 ligand; CD134L; CD252; glycoprotein Gp34; GP34; OX40 antigen ligand; OX40 ligand; OX40L; OX-40L; OX4OL; TAX transcriptionally-activated glycoprotein 1; tax-transcriptionally activated glycoprotein 1 (34kD); tax-transcriptionally activated glycoprotein 1 ligand; TNFSF4; TNLG2B; tumor necrosis factor (ligand) superfamily member 4; tumor necrosis factor (ligand) superfamily, member 4; tumor necrosis factor ligand 2B; tumor necrosis factor ligand superfamily member 4; tumor necrosis factor superfamily member 4; TXGP1; Txgp1l
Common Name OX40L (CD252)
Gene Symbol Tnfsf4
Biological Activity Determined by its ability to bind hOX40 in a functional ELISA.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence QVSHRYPRIQ SIKVQFTEYK KEKGFILTSQ KEDEIMKVQN NSVIINCDGF YLISLKGYFS QEVNISLHYQ KDEEPLFQLK KVRSVNSLMV ASLTYKDKVY LNVTTDNTSL DDFHVNGGEL ILIHQNPGEF CVL
Content And Storage -20°C
Activity Determined by its ability to bind hOX40 in a functional ELISA.
Expression System Insect cells
Form Lyophilized
Purity or Quality Grade ≥ 97% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.