missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Ovastacin Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Brand: R&D Systems AF8549
This item is not returnable.
View return policy
Description
astacin-like metalloendopeptidase Polyclonal specifically detects astacin-like metalloendopeptidase in Human samples. It is validated for Western Blot, Simple Western.
Specifications
| astacin-like metalloendopeptidase | |
| Polyclonal | |
| Western Blot 1:1000 dilution, Simple Western 1:100 dilution | |
| Q6HA08 | |
| ASTL | |
| Mouse myeloma cell line NS0-derived recombinant human Ovastacin Ala24-Asp431 Accession # Q6HA08 | |
| 100 μL | |
| Primary | |
| Detects human Ovasticin in direct ELISAs and Western blots. In direct ELISAs, less then 1% cross-reactivity with recombinant human BMP-1 is observed. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. with Sodium Azide | |
| Astacin-Like Metalloendopeptidase, astacin-like metallo-endopeptidase (M12 family), ASTL, Oocyte Astacin | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 431705 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction