missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Human Ovary Membrane Tissue Lysate (Adult Membrane Tumor)
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
Description
The Human Ovary Membrane Tissue Lysate (Adult Membrane Tumor) is derived from human.
Specifications
Specifications
| Tissue | Reproductive |
| Quantity | 0.1 mg |
| Storage Buffer | HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors |
| Test Specificity | Membrane Protein - Human Tumor Tissue: Ovary |
| Cellular Fraction | Membrane |
| Tissue Condition | Tumor |
| Cross Reactivity | Human |
| Host Species | Human |
| Concentration | 2 mg/mL |
| Product Type | Human Ovary Membrane Tissue Lysate (Adult Tumor) |
| Show More |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction