missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OTP Partial ORF (NP_115485.1, 11 a.a. - 120 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00023440-Q01.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16015345

  • £425.00 / 25µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the homeodomain (HD) family. HD family proteins are helix-turn-helix transcription factors that play key roles in the specification of cell fates. This protein may function during brain development. [provided by RefSeq]

Sequence: RLGMKDAAELLGHREAVKCRLGVGGSDPGGHPGDLAPNSDPVEGATLLPGEDITTVGSTPASLAVSAKDPDKQPGPQGGPNPSQAGQQQGQQKQKRHRTRFTPAQLNELE
Specifications

Specifications

NP_115485.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RLGMKDAAELLGHREAVKCRLGVGGSDPGGHPGDLAPNSDPVEGATLLPGEDITTVGSTPASLAVSAKDPDKQPGPQGGPNPSQAGQQQGQQKQKRHRTRFTPAQLNELE
RUO
OTP
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
23440
OTP (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC3161
OTP
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers