missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Otolin-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8045
This item is not returnable.
View return policy
Description
Otolin-1 Monoclonal specifically detects Otolin-1 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| Otolin-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| A6NHN0 | |
| OTOL1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Otolin-1 Lys24-Pro477 Accession # A6NHN0 | |
| 100 μg | |
| Primary | |
| Detects human Otolin-1 in ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 839418 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C1q and tumor necrosis factor related protein 15, C1qTNF15, LOC131149, OTOL1, otolin 1, otolin-1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 131149 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction