missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Osteopontin/OPN Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems BAF1433
This item is not returnable.
View return policy
Description
Osteopontin/OPN Polyclonal specifically detects Osteopontin/OPN in Human samples. It is validated for Western Blot, Immunocytochemistry, ELISA Detection (Matched Antibody Pair).
Specifications
| Osteopontin/OPN | |
| Polyclonal | |
| Biotin | |
| BNSP, Bone sialoprotein 1, Eta-1, MGC110940, Nephropontin, OPN, osteopontin, secreted phosphoprotein 1bone sialoprotein I, early T-lymphocyteactivation 1), secreted phosphoprotein-1 (osteopontin, bone sialoprotein), Spp1, SPP-1, SPP1/CALPHA1 fusion, Urinary stone protein, uropontin | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6696 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry, ELISA | |
| LYOPH | |
| Western Blot 0.1 μg/mL, Immunocytochemistry 1.7-15 μg/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 μg/mL | |
| SPP1 | |
| Human milk-derived Osteopontin/OPN | |
| 50 μg | |
| Primary | |
| Detects human Osteopontin/OPN in ELISAs and Western blots. In sandwich ELISAs, less than 0.2% cross-reactivity with recombinant mouse (rm) Osteopontin and bovine Osteopontin is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction