missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Osteocalcin Instant ELISA™ Kit

Product Code. 15571507
Click to view available options
Quantity:
128 Tests
Unit Size:
128 tests
This item is not returnable. View return policy

Product Code. 15571507

Brand: Invitrogen™ BMS2020INST

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

The Human Osteocalcin ELISA quantitates Hu Osteocalcin in human serum, plasma, cell culture supernatants, urine, amniotic fluid, bile, or other body fluids. The assay will exclusively recognize both natural and recombinant Hu Osteocalcin. Principle of the method The Human Osteocalcin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the binding of the second (detector) antibody to the target on a different epitope from the capture antibody. An antibody conjugated with enzyme binds the formed sandwich. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Osteocalcin belongs to the osteocalcin/matrix Gla-protein family and constitutes 1-2% of the total bone protein. It binds strongly to apatite and calcium. Gamma-carboxyglutamate residues are formed by vitamin K dependent carboxylation. These residues are essential for the binding of calcium.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P02818
Assay Range 1.2 to 75 ng/mL
Assay Sensitivity 0.2 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Application) ELISA
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 632
Gene Symbol BGP,OC,OCN
Interassay CV 8.1%
Intraassay CV 8.3
Kit Contents Pre-coated 96 well Microplate, Detection Antibody, Anti-IgG-HRP, Sample Diluent, lyophilized, Standard (8-well strips x 4), Wash Buffer Concentrate, Substrate Solution, Sample Diluent, Stop Solution, Lysis Buffer 10x, Adhesive Plate Covers
Label or Dye HRP
Quantity 128 Tests
Regulatory Status RUO
Sample Volume Plasma 25 μL, Serum 25 μL, Supernatant 25 μL
Storage Requirements -20°C
Synonym Gamma-carboxyglutamic acid-containing protein
Target Osteocalcin
Test Time 15 mins
Total Assay Time 2 hrs 10 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.