missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ORP150/HSP12A Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF5558-SP
This item is not returnable.
View return policy
Description
ORP150/HSP12A Polyclonal specifically detects ORP150/HSP12A in Human samples. It is validated for Western Blot.
Specifications
| ORP150/HSP12A | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 150 kDa oxygen-regulated protein, 170 kDa glucose-regulated protein, DKFZp686N08236, FLJ97572, glucose-regulated protein 170, GRP170, GRP-170, HSP12A, HYOU1, hypoxia up-regulated 1, hypoxia up-regulated protein 1, ORP-150, ORP150FLJ94899, oxygen regulated protein (150kD) | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 10525 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9Y4L1 | |
| HYOU1 | |
| Mouse myeloma cell line NS0-derived recombinant human ORP150/HSP12A Met695-Leu994 Accession # Q9Y4L1 | |
| 25 μg | |
| Primary | |
| Detects human ORP150/HSP12A in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) HSP-70, rhHSPA-2, and rhHSPH-1 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction