missing translation for 'onlineSavingsMsg'
Learn More

Human Orexin R2/HCRTR2 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Product Code. 18746749
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18746749 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18746749 Supplier R&D Systems Supplier No. IC5246G100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Orexin R2/HCRTR2 Monoclonal specifically detects Orexin R2/HCRTR2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Orexin R2/HCRTR2
Applications Flow Cytometry
Classification Monoclonal
Clone 456738
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 488
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. AAC39602
Gene Alias HCRTR2, hypocretin (orexin) receptor 2, Hypocretin receptor type 2, Orexin Receptor 2, orexin receptor type 2, OrexinR2, Ox-2-R, Ox2-R, OX2R
Gene Symbols HCRTR2
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Orexin R2/HCRTR2 Met1-Trp444 Accession # AAC39602
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3062
Test Specificity Detects human Orexin R2/HCRTR2. Stains human Orexin R2/HCRTR2 transfectants but not irrelevant transfectants.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.