missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Orexin (aa 38-68) Control Fragment Recombinant Protein

Product Code. 30207898
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30207898

Brand: Invitrogen™ RP109072

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Neuropeptides that play a significant role in the regulation of food intake and sleep-wakefulness, possibly by coordinating the complex behavioral and physiologic responses of these complementary homeostatic functions. A broader role in the homeostatic regulation of energy metabolism, autonomic function, hormonal balance and the regulation of body fluids, is also suggested. Orexin-A binds to both OX1R and OX2R with a high affinity, whereas orexin-B binds only to OX2R with a similar high affinity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O43612
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 3060
Name Human Orexin (aa 38-68) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias HCRT; Hcrt1; Hcrt2; Hypocretin; hypocretin (orexin) neuropeptide; hypocretin neuropeptide precursor; Hypocretin-1; Hypocretin-2; NRCLP1; Orexin; orexin A; orexin B; Orexin-A; Orexin-B; OX; PPORX; Ppox; prepro-orexin
Common Name Orexin
Gene Symbol HCRT
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DCCRQKTCSCRLYELLHGAGNHAAGILTLGK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.