missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human OR1Q1 (aa 1-25) Control Fragment Recombinant Protein

Product Code. 30202495
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30202495

Brand: Invitrogen™ RP100539

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (88%), Rat (88%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60842 (PA5-60842. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q15612
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 158131
Name Human OR1Q1 (aa 1-25) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias HSTPCR106; Olfactory receptor 1Q1; olfactory receptor 1Q2; Olfactory receptor 1Q3; olfactory receptor 9-A; olfactory receptor family 1 subfamily Q member 1; olfactory receptor OR9-25; Olfactory receptor TPCR106; olfactory receptor, family 1, subfamily Q, member 1; olfactory receptor, family 1, subfamily Q, member 2; olfactory receptor, family 1, subfamily Q, member 3; OR1Q; OR1Q1; OR1Q2; OR1Q3; OR9-25; OR9-A; OST226; OST226OR9-A; TPCR106
Common Name OR1Q1
Gene Symbol OR1Q1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MDNSNWTSVSHFVLLGISTHPEEQI
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis