missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Opticin Antibody, R&D Systems™

Goat Polyclonal Antibody
£167.00 - £432.00
Specifications
| Antigen | Opticin |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18727197
![]() |
R&D Systems
AF3148-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18091492
|
R&D Systems
AF3148 |
100 μg |
£432.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Opticin Polyclonal specifically detects Opticin in Human samples. It is validated for Western Blot.Specifications
| Opticin | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| oculoglycan, Oculogylcan, OPT, OPTC, opticin | |
| OPTC | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q9UBM4 | |
| 26254 | |
| Mouse myeloma cell line NS0-derived recombinant human Opticin Ala20-Thr332 Accession # Q9UBM4 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Opticin in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant mouse Opticin is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title