missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human OPA3 (aa 103-168) Control Fragment Recombinant Protein

Product Code. 30182331
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30182331

Brand: Invitrogen™ RP99016

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (59%), Rat (59%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60855 (PA5-60855. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The mouse ortholog of this protein co-purifies with the mitochondrial inner membrane. Mutations in this gene have been shown to result in 3-methylglutaconic aciduria type III and autosomal dominant optic atrophy and cataract. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9H6K4
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 80207
Name Human OPA3 (aa 103-168) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias D630048P19Rik; FLJ22187; FLJ25932; Gm1425; Gm472; MGA3; MGC75494; OPA3; OPA3 outer mitochondrial membrane lipid metabolism regulator; optic atrophy 3; optic atrophy 3 (autosomal recessive, with chorea and spastic paraplegia); Optic atrophy 3 (Iraqi-Jewish'optic atrophy plus'); optic atrophy 3 protein; Optic atrophy 3 protein homolog; outer mitochondrial membrane lipid metabolism regulator OPA3
Common Name OPA3
Gene Symbol OPA3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence RHQLQQRRKEKERRVAREALRGEVGHLGLALEELQAQVQATSTQLALEELRAQLQEVRAHLCLRDP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado