missing translation for 'onlineSavingsMsg'
Learn More

Human Oncostatin M/OSM Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 8 publications

£178.00 - £1765.00

Specifications

Antigen Oncostatin M/OSM
Clone 17001
Dilution Western Blot 1 ug/mL, Neutralization 0.5-2.0 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Applications Western Blot, Neutralization, ELISA
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18773348
GREENER_CHOICE
View Documents
R&D Systems
MAB295-SP
25 μg
£178.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18739437
GREENER_CHOICE
View Documents
R&D Systems
MAB295-100
100 μg
£447.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18722597
GREENER_CHOICE
View Documents
R&D Systems
MAB295-500
500 μg
£1765.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Oncostatin M/OSM Monoclonal specifically detects Oncostatin M/OSM in Human samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications

Specifications

Oncostatin M/OSM
Western Blot 1 ug/mL, Neutralization 0.5-2.0 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Monoclonal
Mouse
Human
P13725
5008
E. coli-derived recombinant human Oncostatin M/OSM Ala26-Arg234 Accession # P13725
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human Oncostatin M in ELISAs and Western blots. In ELISAs, this antibody does not cross-react with recombinant human (rh) IL-6, rhIL-11, rhCNTF, or rhLIF.
17001
Western Blot, Neutralization, ELISA
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
MGC20461, oncostatin M, oncostatin-M
OSM
IgG2a
Protein A or G purified from ascites
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.