missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Oncostatin M/OSM Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC2951S-100UG
This item is not returnable.
View return policy
Description
Oncostatin M/OSM Monoclonal specifically detects Oncostatin M/OSM in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Oncostatin M/OSM | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P13725 | |
| OSM | |
| E. coli-derived recombinant human Oncostatin M/OSM Ala26-Arg221 Accession # P13725 | |
| RUO | |
| 5008 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 17022 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| MGC20461, oncostatin M, oncostatin-M | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Oncostatin M/OSM in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Oncostatin M, recombinant human (rh) CLC, rhCNTF, rhCardiotrophin-1, rhIL-6, rhIL-11, or rhLIF is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction