missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human OMgp Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1673-SP
This item is not returnable.
View return policy
Description
OMgp Polyclonal specifically detects OMgp in Human samples. It is validated for Western Blot.
Specifications
| OMgp | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| oligodendrocyte myelin glycoprotein, OMG, OMGPoligodendrocyte-myelin glycoprotein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4974 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| P23515 | |
| OMG | |
| Mouse myeloma cell line NS0-derived recombinant human OMgp Ile25-Asn420 Accession # P23515 | |
| 25 μg | |
| Primary | |
| Detects human OMgp in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 70% cross-reactivity with recombinant mouse OMgp is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction