missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Olig1 Antibody, R&D Systems™
Goat Polyclonal Antibody
£464.80
Specifications
Antigen | Olig1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Description
Olig1 Polyclonal specifically detects Olig1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Olig1 | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Human | |
bHLHb6, BHLHE21, Class B basic helix-loop-helix protein 6, Class E basic helix-loop-helix protein 21, helix-loop-helix protein, class B, 6, oligo1, oligodendrocyte lineage transcription factor 1, oligodendrocyte transcription factor 1, oligodendrocyte-specific bHLH transcription factor 1 | |
OLIG1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
BAD93028 | |
116448 | |
E. coli-derived recombinant human Olig1 Met12-Lys283 Accession # BAD93028 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Olig1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows less than 5% cross-reactivity with recombinant human (rh) Olig2 and rhOlig3. |
For Research Use Only