missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human ODC1 (NP_002530.1) VersaClone cDNA

Product Code. 18720379
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18720379 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18720379 Supplier R&D Systems™ Supplier No. RDC3041
  • ONLINE EXCLUSIVE PRICE
    £307.00
    £289.80 / 10µg
    Save £17.20
    5% Off
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Contains gene inserts that are fully sequenced and produced with high quality

TRUSTED_SUSTAINABILITY

Specifications

Gene Ornithine Decarboxylase
Accession Number NP_002530.1
Gene ID (Entrez) 4953
Gene Alias ODC, ODCEC 4.1.1.17, ornithine decarboxylase, ornithine decarboxylase 1
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.