missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human OCIL/CLEC2d Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3480
This item is not returnable.
View return policy
Description
OCIL/CLEC2d Polyclonal specifically detects OCIL/CLEC2d in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| OCIL/CLEC2d | |
| Polyclonal | |
| Unconjugated | |
| Q9UHP7 | |
| CLEC2D | |
| E. coli-derived recombinant human OCIL/CLEC2d Ser57-Val191 Accession # Q9UHP7 | |
| 100 μg | |
| Primary | |
| Detects human OCIL/CLEC2d in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant mouse (rm) OCIL is observed and less than 1% cross-reactivity with recombinant human (rh) CLEC2B, and rhCLEC2A is observed. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| CLAXLLT-1, CLEC2D, C-type lectin domain family 2 member D, C-type lectin domain family 2, member D, C-type lectin related f, Lectin-like NK cell receptor, Lectin-like transcript 1, LLT1, LLT1Osteoclast inhibitory lectin, OCILmember D | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 29121 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction