missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human OCIL/CLEC2d Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3480-SP
This item is not returnable.
View return policy
Description
OCIL/CLEC2d Polyclonal specifically detects OCIL/CLEC2d in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| OCIL/CLEC2d | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CLAXLLT-1, CLEC2D, C-type lectin domain family 2 member D, C-type lectin domain family 2, member D, C-type lectin related f, Lectin-like NK cell receptor, Lectin-like transcript 1, LLT1, LLT1Osteoclast inhibitory lectin, OCILmember D | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 29121 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Q9UHP7 | |
| CLEC2D | |
| E. coli-derived recombinant human OCIL/CLEC2d Ser57-Val191 Accession # Q9UHP7 | |
| 25 μg | |
| Primary | |
| Detects human OCIL/CLEC2d in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant mouse (rm) OCIL is observed and less than 1% cross-reactivity with recombinant human (rh) CLEC2B, and rhCLEC2A is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction