missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems Human Obesity Premixed Magnetic Luminex™ Performance Assay Kit

Product Code. 30002106
Change view
Click to view available options
Quantity:
2 Plex
4 Plex
5 Plex
6 Plex
7 Plex
8 Plex
9 Plex
Unit Size:
1 set
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30002106 8 Plex 1 set
30002112 9 Plex 1 set
30002100 7 Plex 1 set
30002092 6 Plex 1 set
30002087 5 Plex Each
30002082 4 Plex Each
30002073 2 Plex 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 30002106 Supplier R&D Systems Supplier No. FCSTM0808

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 8 Plex
Species Human
Binding Target Obesity Assay Kits
For Use With (Application) ELISA
Content And Storage Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.