missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human OBCAM/OPCML Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB27771-SP
This item is not returnable.
View return policy
Description
OBCAM/OPCML Monoclonal specifically detects OBCAM/OPCML in Human samples. It is validated for Western Blot.
Specifications
| OBCAM/OPCML | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q14982 | |
| OPCML | |
| Mouse myeloma cell line NS0-derived recombinant human OBCAM Gly28-Asn322 Accession # Q14982 | |
| 25 μg | |
| Primary | |
| Detects human OBCAM on direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) ALCAM, rhBCAM, rhEpCAM, rhNCAM, recombinant mouse (rm) MAdCAM, rmOCAM, or rmNCAM-L1. | |
| Human | |
| Purified |
| Western Blot | |
| 341723 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IgLON family member 1, IGLON1opioid-binding protein/cell adhesion molecule-like, OBCAMopiate binding-cell adhesion molecule, OPCM, OPCML, opioid binding protein/cell adhesion molecule-like, opioid binding protein/cell adhesion molecule-like preprotein, Opioid-binding cell adhesion molecule, opioid-binding protein/cell adhesion molecule | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4978 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction