missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human OATP1 (aa 265-315) Control Fragment Recombinant Protein

Product Code. 30202031
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30202031

Brand: Invitrogen™ RP105520

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (65%), Rat (65%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-67149 (PA5-67149. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The organic anion transporting polypeptides, OATP-A (also designated OATP1, OATP1A2 and SLC21A3) and OATP-C (also designated OATP2, SLC21A6 and LST1), mediate hepatic uptake of cardiac glycosides. The expression of OATP-C, but not OATP-A, is inducible by phenobarbital and pregnenolone-16a-carbonitrile, resulting in the increased capacity of the liver to extract cardiac glycosides from the plasma. OATP-A is expressed in liver and kidney and helps mediate sodium-independent uptake of the anionic steroid conjugates dehydroepiandrosterone sulfate, estradiol-17-glucuronide and prostaglandin. OATP-C is exclusively expressed in liver and is localized to the basolateral hepatocyte membrane.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P46721
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6579
Name Human OATP1 (aa 265-315) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias A530084B21; Brain digoxin carrier protein; brain-specific organic anion transporter; membrane transporter; OATP; Oatp1; OATP-1; Oatp1a1; OATP1A2; Oatp1a4; Oatp2; OATPA; OATP-A; OATP-B1; organic anion transporter 2; organic anion transporting polypeptide 1; organic anion transporting polypeptide 1a2; organic anion transporting polypeptide 1a2 variant 1; organic anion transporting polypeptide A; organic anion-transporting polypeptide 1; Slc21a1; SLC21A3; Slc21a5; Slco1a1; SLCO1A2; Slco1a4; SO1A2; Sodium-independent organic anion transporter; sodium-independent organic anion transporter 1; Sodium-independent organic anion-transporting polypeptide 1; sodium-independent organic anion-transporting polypeptide 2; solute carrier family (organic anion transporter) member 1; solute carrier family (organic anion transporter) member 3; solute carrier family 21 (organic anion transporter), member 1; solute carrier family 21 (organic anion transporter), member 3; solute carrier family 21 (organic anion transporter), member 5; solute carrier family 21 member 1; Solute carrier family 21 member 3; solute carrier family 21 member 5; solute carrier family 21, member 1; solute carrier organic anion transporter family member 1A1; Solute carrier organic anion transporter family member 1A2; solute carrier organic anion transporter family member 1A4; solute carrier organic anion transporter family, member 1a1; solute carrier organic anion transporter family, member 1A2; solute carrier organic anion transporter family, member 1a4
Common Name OATP1
Gene Symbol SLCO1A2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence FLPNTLPKEGLETNADIIKNENEDKQKEEVKKEKYGITKDFLPFMKSLSCN
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.