missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human OAS2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1925-SP
This item is not returnable.
View return policy
Description
OAS2 Monoclonal specifically detects OAS2 in Human samples. It is validated for Immunoprecipitation, Immunocytochemistry.
Specifications
| OAS2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| OAS2 | |
| 2Æ,5Æ-Oligoadenylate Synthetase-2 | |
| 25 μg | |
| Primary | |
| Detects human OAS2. | |
| Human | |
| Purified |
| Immunoprecipitation, Immunocytochemistry | |
| HLS56/3 | |
| Immunoprecipitation, Immunocytochemistry 8-25 ug/mL | |
| (2-5')oligo(A) synthase 2, (2'-5')oligo(A) synthetase 2, (2-5')oligo(A) synthetase 2, 2'5' oligoadenylate synthetase 2, 2-5A synthase 2, 2-5A synthetase 2, 2'-5'-oligoadenylate synthase 2, 2'-5'-oligoadenylate synthetase 2 (69-71 kD), 2'-5'-oligoadenylate synthetase 2, 69/71kDa, EC 2.7.7, EC 2.7.7.-, MGC78578, p69 OAS p71 OAS, p69OAS p71OAS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4939 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction