missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Nyctalopin/NYX (NP_072089) VersaClone cDNA
Description
- Also known as: CLRP; NBM1; CSNB1; CSNB4; CSNB1A
- NYX belongs to the small leucinerich proteoglycan (SLRP) family of proteins.
- Defects in NYX are the cause of congenital stationary night blindness type 1 (CSNB1), also called X-linked congenital stationary night blindness (XLCSNB).
- The role of other SLRP proteins suggests that mutations in NYX disrupt developing retinal interconnections involving the ON-bipolar cells, leading to the visual losses seen in patients with complete CSNB.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hNYX |
| Accession Number | NP_072089 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1459 bp |
| Gene ID (Entrez) | 60506 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?