missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human NXT2 (aa 3-61) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (34%), Rat (34%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-67222 (PA5-67222. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Regulator of protein export for NES-containing proteins. Also plays a role in mRNA nuclear export.
Specifications
Specifications
| Accession Number | Q9NPJ8 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 55916 |
| Name | Human NXT2 (aa 3-61) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 6330587F24Rik; AI787442; BM-025; DC9; NTF2-related export protein 2; nuclear transport factor 2 like export factor 2; nuclear transport factor 2-like export factor 2; NXT2; P15-2; protein p15-2; RGD1560204 |
| Common Name | NXT2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction