missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NUP35 (aa 89-157) Control Fragment Recombinant Protein

Product Code. 30205109
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30205109

Brand: Invitrogen™ RP91855

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (88%), Rat (88%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-110837 (PA5-110837. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Nucleoporin Nup35 or Mitotic phosphoprotein 44 is a nuclear protein known to be present on nuclear pore complex and is tightly associated with the nuclear membrane and lamina. This RNA associated protein belongs to the Nup53 family and has a MPPN (mitotic phosphoprotein N' end) domain. It has a small number of scattered phenylalanine-glycine repeats, but not the large domains seen in other nucleoporins. Though the exact function of NUP35 is yet to be revealed, it may function as a component of the nuclear pore complex (NPC) which are collectively referred to as nucleoporins (NUPs). It also may play the role of both NPC structural components and of docking or interaction partners for transiently associated nuclear transport factors, and may also play an important part in the association of MAD1, a transcriptional repressor with the NPC. NUP35 is widely expressed in most of the tissues.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8NFH5
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 129401
Name Human NUP35 (aa 89-157) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2310006I24Rik; 35 kDa nucleoporin; 35 kDa; 5330402E05Rik; Mitotic phosphoprotein 44; Mp44; MP-44; NO44; NP44; Nuclear pore complex protein Nup53; nucleoporin 35; nucleoporin 35 kDa; nucleoporin NUP35; Nucleoporin NUP53; Nup35; NUP53
Common Name NUP35
Gene Symbol NUP35
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PPVRSIYDDISSPGLGSTPLTSRRQPNISVMQSPLVGVTSTPGTGQSMFSPASIGQPRKTTLSPAQLDP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.