missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Numb Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4338-SP
This item is not returnable.
View return policy
Description
Numb Monoclonal specifically detects Numb in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| Numb | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| AAH68476 | |
| NUMB | |
| E. coli-derived recombinant human Numb Thr356-Leu592 Accession # AAH68476 | |
| 25 μg | |
| Primary | |
| Detects human Numb. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 447716 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| h-Numb, mnb, m-numb, numb (Drosophila) homolog, numb homolog (Drosophila), protein numb homolog, Protein S171, S171 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8650 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering