missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NUDT4 (aa 127-180) Control Fragment Recombinant Protein

Product Code. 30209239
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30209239

Brand: Invitrogen™ RP91860

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53612 (PA5-53612. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Cleaves a beta-phosphate from the diphosphate groups in PP-InsP5 (diphosphoinositol pentakisphosphate), PP-InsP4 and [PP]2-InsP4 (bisdiphosphoinositol tetrakisphosphate), suggesting that it may play a role in signal transduction. Also able to catalyze the hydrolysis of dinucleoside oligophosphate Ap6A, but not Ap5A. The major reaction products are ADP and p4a from Ap6A. Also able to hydrolyze 5-phosphoribose 1-diphosphate. Does not play a role in U8 snoRNA decapping activity. Binds U8 snoRNA.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NZJ9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 11163
Name Human NUDT4 (aa 127-180) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 4933436C10Rik; Diadenosine 5',5'''-P1,P6-hexaphosphate hydrolase 2; diphosphoinositol polyphosphate phosphohydolase type II; diphosphoinositol polyphosphate phosphohydrolase 2; Diphosphoinositol polyphosphate phosphohydrolase NUDT4B; diphosphoinositol polyphosphate phosphohydrolase type 2; DIPP2; DIPP-2; DIPP2alpha; DIPP-2 B; DIPP2beta; DRCF-5; HDCMB47P; KIAA0487; Nucleoside diphosphate-linked moiety x motif 4; Nucleoside diphosphate-linked moiety x motif 4 B; nudix (nucleoside diphosphate linked moiety X)-type motif 4; nudix hydrolase 4; Nudix hydrolase 4 B; Nudix motif 4; Nudix motif 4 B; nudix-type motif 4; NUDT4; NUDT4B; rDIPP2
Common Name NUDT4
Gene Symbol NUDT4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KVLQCHKPVHAEYLEKLKLGCSPANGNSTVPSLPDNNALFVTAAQTSGLPSSVR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.