Learn More
Abcam Human NUDT2 knockout HeLa cell line
Product Code. 32157877 Shop All Abcam Products
Change view
Click to view available options
Quantity:
2 x 1,000,000 Cells
Unit Size:
Pack of 2
Product Code. Quantity Unit Size
32157877 2 x 1,000,000 Cells Pack of 2
1 options
Product Code. 32157877 Supplier Abcam Supplier No. ab2657522x1000000Cells_
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
NUDT2 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 16 bp insertion in exon 2 and 1 bp deletion in exon 2 and 8 bp deletion in exon 2. To order both knockout and wild-type control cells: select 2 x 1000000 Cells...
NUDT2 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 16 bp insertion in exon 2 and 1 bp deletion in exon 2 and 8 bp deletion in exon 2. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 2 x 1,000,000 Cells