missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NUDT10 (aa 129-154) Control Fragment Recombinant Protein

Product Code. 30196894
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30196894 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30196894 Supplier Invitrogen™ Supplier No. RP104155

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63467 (PA5-63467. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Cleaves a beta-phosphate from the diphosphate groups in PP-InsP5 (diphosphoinositol pentakisphosphate), suggesting that it may play a role in signal transduction. Also able to catalyze the hydrolysis of dinucleoside oligophosphates, with Ap6A and Ap5A being the preferred substrates. The major reaction products are ADP and p4a from Ap6A and ADP and ATP from Ap5A. Also able to hydrolyze 5-phosphoribose 1-diphosphate.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8NFP7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 170685
Name Human NUDT10 (aa 129-154) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AI853080; APS2; Diadenosine 5',5'''-P1,P6-hexaphosphate hydrolase 3-alpha; Diadenosine 5',5'''-P1,P6-hexaphosphate hydrolase 3-beta; Diadenosine hexaphosphate hydrolase (AMP-forming); diphosphoinositol polyphosphate phosphohydrolase 3-alpha; Diphosphoinositol polyphosphate phosphohydrolase 3-beta; DIPP3 alpha; Dipp3a; DIPP-3-alpha; DIPP3-alpha; Dipp3b; DIPP-3-beta; hAps2; hDIPP3alpha; MNCb-1696; Nucleoside diphosphate-linked moiety x motif 10; Nucleoside diphosphate-linked moiety x motif 11; nudix (nucleoside diphosphate linked moiety X)-type motif 10; nudix hydrolase 10; nudix motif 10; Nudix motif 11; Nudt10; Nudt11; RGD1564763
Common Name NUDT10
Gene Symbol NUDT10
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LQCHKPVHAEYLEKLKLGGSPTNGNS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.