missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Nudel (aa 42-117) Control Fragment Recombinant Protein

Product Code. 30205714
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30205714

Brand: Invitrogen™ RP92320

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53669 (PA5-53669. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a thiol-activated peptidase that is phosphorylated in M phase of the cell cycle. Phosphorylation regulates the cell cycle-dependent distribution of this protein, with a fraction of the protein bound strongly to centrosomes in interphase and localized to mitotic spindles in early M phase. Overall, this protein plays a role in nervous system development. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9GZM8
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 81565
Name Human Nudel (aa 42-117) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2600006O07Rik; ATP-sensitive inward rectifier potassium channel 15; DKFZp451M0318; endooligopeptidase A; EOPA; Inward rectifier K(+) channel Kir4.2; Kcnj15; Kir4.2; LIS1-interacting protein NUDEL; LIS1-interacting protein NUDEL; endooligopeptidase A; MITAP1; Mitosin-associated protein 1; mitosin-associated protein MITAP1; mNudel; mNudE-L; NDE1L1; NDE2; Ndel1; nuclear disribution gene E like; nuclear distribution gene E-like homolog 1; nuclear distribution gene E-like homolog 1 (A. nidulans); nuclear distribution protein nudE-like 1; nudE neurodevelopment protein 1 like 1; nudE neurodevelopment protein 1-like 1; nudE nuclear distribution E homolog-like 1; nudE nuclear distribution gene E homolog like 1; nudE nuclear distribution gene E homolog-like 1; Nude2; NUDEL; NUDE-like protein; Potassium channel, inwardly rectifying subfamily J member 15; potassium channel, inwardly rectifying subfamily J, member 15; potassium inwardly-rectifying channel, subfamily J, member 15; Protein mNudE-like; Protein Nudel; unnamed protein product
Common Name Nudel
Gene Symbol Ndel1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EGSRELEAELEAQLVQAEQRNRDLQADNQRLKYEVEALKEKLEHQYAQSYKQVSVLEDDLSQTRAIKEQLHKYVRE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.