missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NTH1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2675
This item is not returnable.
View return policy
Description
NTH1 Polyclonal specifically detects NTH1 in Human samples. It is validated for Western Blot.
Specifications
| NTH1 | |
| Polyclonal | |
| Unconjugated | |
| P78549 | |
| NTHL1 | |
| E. coli-derived recombinant human NTH1 Met1-Leu312 Accession # P78549 | |
| 100 μg | |
| Primary | |
| Detects human NTH1 in Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| EC 4.2.99.18, endonuclease III-like protein 1, nth endonuclease III-like 1 (E. coli), NTHL1, OCTS3, OCTS3NTH1nth (E.coli endonuclease III)-like 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4913 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction