missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NTB-A/SLAMF6 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
£188.00 - £498.00
Specifications
| Antigen | NTB-A/SLAMF6/CD352 |
|---|---|
| Clone | 292811 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18778507
![]() |
R&D Systems
FAB19081A-025 |
25 Tests |
£188.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18749336
![]() |
R&D Systems
FAB19081A-100 |
100 Tests |
£498.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NTB-A/SLAMF6/CD352 Monoclonal specifically detects NTB-A/SLAMF6/CD352 in Human samples. It is validated for Flow Cytometry.Specifications
| NTB-A/SLAMF6/CD352 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Activating NK receptor, CD352, CD352 antigen, KALIFLJ50657, Ly108, NK-T-B-antigen, NTBA, NTBA receptor, NTB-AMGC104953, NTBAT- and B-cell antigen, SF2000, SLAM family member 6, SLAMF6 | |
| SLAMF6 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human NTB-A/SLAMF6 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) BLAME or rhCRACC is observed. |
| 292811 | |
| Flow Cytometry | |
| APC | |
| Mouse | |
| Human | |
| Q96DU3 | |
| 114836 | |
| Mouse myeloma cell line NS0-derived recombinant human NTB-A/SLAMF6 Leu28-Lys225 Accession # Q96DU3 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title