missing translation for 'onlineSavingsMsg'
Learn More

Human NT-4 Antibody, R&D Systems™

Product Code. 18199930
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18199930 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18199930 Supplier R&D Systems Supplier No. MAB268

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

NT-4 Monoclonal specifically detects NT-4 in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NT-4
Applications Western Blot, ELISA
Classification Monoclonal
Clone 36526
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias GLC10, GLC1O, neurotrophic factor 4, neurotrophic factor 5, neurotrophin 4, neurotrophin 5 (neurotrophin 4/5), neurotrophin-4, neurotrophin-5, Neutrophic factor 4, NT4, NT-4, NT-4/5, NT-5, NTF4, NTF5, NTF5NT5
Gene Symbols NTF4
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human NT-4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4909
Test Specificity Detects human NT-4 in direct ELISAs and Western blots. In Western blots, this antibody shows less than 1% cross-reactivity with recombinant human (rh) NT-3 and rhBDNF.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.