missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human NT-4, Animal-Free Recombinant Protein, PeproTech®

Product Code. 17813093
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
This item is not returnable. View return policy

Product Code. 17813093

Brand: Gibco™ AF45004100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human NT-4 is a noncovalently linked homodimer of two 14.0 kDa polypeptide monomers (260 total amino acid residues). This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This gene is a member of a family of neurotrophic factors, neurotrophins, that control survival and differentiation of mammalian neurons. The expression of this gene is ubiquitous and less influenced by environmental signals. While knock-outs of other neurotrophins including nerve growth factor, brain-derived neurotrophic factor, and neurotrophin 3 prove lethal during early postnatal development, NTF5-deficient mice only show minor cellular deficits and develop normally to adulthood.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P34130
For Use With (Application) ELISA, Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 4909
Molecular Weight (g/mol) 28.1 kDa
Name Human NT-4, Animal-Free
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias 2900040K06Rik; AI462899; GLC10; GLC1O; neurotrophic factor 4; neurotrophic factor 5; neurotrophin 4; Neurotrophin 4/5; neurotrophin 5; Neurotrophin 5 (neurotrophin 4/5); neurotrophin 5 (neurotrophin 4/5); neurotrophin-4; Neurotrophin-5; neutrophic factor 4; Neutrophic factor 5; NT4; NT-4; NT4/5; NT-4/5; NT4/NT5; NT4P; NT5; NT-5; Ntf4; Ntf5; Ntf-5
Common Name NTF4
Gene Symbol NTF4
Biological Activity Determined by its ability to bind recombinant human TrkB Fc Chimera in a functional ELISA.
Conjugate Unconjugated
Recombinant Recombinant
Sequence GVSETAPASR RGELAVCDAV SGWVTDRRTA VDLRGREVEV LGEVPAAGGS PLRQYFFETR CKADNAEEGG PGAGGGGCRG VDRRHWVSEC KAKQSYVRAL TADAQGRVGW RWIRIDTACV CTLLSRTGRA
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.