missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NSUN5 (aa 133-210) Control Fragment Recombinant Protein

Product Code. 30210082
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30210082

Brand: Invitrogen™ RP91740

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (88%), Rat (88%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54228 (PA5-54228. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

NSUN5 is a member of the evolutionarily conserved NOL1/NOP2/Sun domain family. The encoded protein may function as a DNA methyltransferase in the nucleus. The gene is deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.23. Alternative splicing of this gene results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96P11
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 55695
Name Human NSUN5 (aa 133-210) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 9830109N13Rik; AI326939; FLJ10267; MGC986; NOL1; NOL1/NOP2/Sun domain family member 5; NOL1/NOP2/Sun domain family, member 5; NOL1R; NOL1-related protein; NOP2/Sun domain family, member 5; NOP2/Sun domain family, member 5 A; NOP2/Sun RNA methyltransferase family member 5; Nsun5; NSUN5A; p120; p120(NOL1); Probable 28 S rRNA (cytosine-C(5))-methyltransferase; putative methyltransferase NSUN5; RGD1309268; WBSCR20; Wbscr20a; Williams Beuren syndrome chromosome region 20 A; Williams Beuren syndrome chromosome region 20 A homolog; Williams-Beuren syndrome chromosomal region 20 A protein; Williams-Beuren syndrome chromosomal region 20 A protein homolog; Williams-Beuren syndrome critical region protein 20; Williams-Beuren syndrome critical region protein 20 copy A
Common Name NSUN5
Gene Symbol Nsun5
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence QLPRFVRVNTLKTCSDDVVDYFKRQGFSYQGRASSLDDLRALKGKHFLLDPLMPELLVFPAQTDLHEHPLYRAGHLIL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.