missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NSE2 (aa 121-163) Control Fragment Recombinant Protein

Product Code. 30203095
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30203095

Brand: Invitrogen™ RP104203

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (79%), Rat (79%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-64243 (PA5-64243. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes the large subunit of DNA damage-binding protein which is a heterodimer composed of a large and a small subunit. This protein functions in nucleotide-excision repair. Its defective activity causes the repair defect in the patients with xeroderma pigmentosum complementation group E (XPE). However, it remains for mutation analysis to demonstrate whether the defect in XPE patients is in this gene or the gene encoding the small subunit. In addition, Best vitelliform mascular dystrophy is mapped to the same region as this gene on 11q, but no sequence alternations of this gene are demonstrated in Best disease patients.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96MF7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 286053
Name Human NSE2 (aa 121-163) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1110014D18Rik; AI661537; C8orf36; E3 SUMO-protein ligase NSE2; E3 SUMO-protein transferase NSE2; FLJ32440; hMMS21; methyl methanesulfonate sensitivity gene 21; MMS21; MMS21 homolog; non-SMC element 2 homolog; non-SMC element 2 homolog (MMS21, S. cerevisiae); non-SMC element 2, MMS21 homolog; non-SMC element 2, MMS21 homolog (S. cerevisiae); non-structural maintenance of chromosomes element 2 homolog; NSE2; NSE2 (MMS21) homolog, SMC5-SMC6 complex SUMO ligase; NSE2/MMS21 homolog, SMC5-SMC6 complex SUMO ligase; NSMCE2; RGD1305156; Unknown (protein for MGC:133996); zinc finger, MIZ-type containing 7; ZMIZ7
Common Name NSE2
Gene Symbol Nsmce2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence QNNEKFVQFKQQLKELKKQCGLQADREADGTEGVDEDIIVTQS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.