missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human NRF2 ELISA Kit
Description
Human NRF2 quantitates human NRF2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human NRF2.
Nfr1 (also designated TCF11), Nrf2 and Nfr3 are bZIP transcription factors that heterodimer with Maf proteins to bind MARE (Maf recognition elements) sequences. Nrf2 is widely expressed and is thought to translocate to the nucleus after treatment with xenobiotics and antioxidants, which stimulate its release from a repressor protein Keap1.
Specifications
Specifications
| Accession Number | AI194320; E4TF1-60; E4TF1A; HEBP1; NFE2L2; NF-E2-related factor 2; NFE2-related factor 2; NFT2; NRF2; NRF2A; nuclear factor erythroid 2-like 2; nuclear factor erythroid 2-related factor 2; nuclear factor erythroid-derived 2-like 2; nuclear factor, erythroid 2 like 2; nuclear factor, erythroid 2-like 2; nuclear factor, erythroid derived 2, like 2 |
| Assay Range | 14 to 10,000 pg/mL |
| Assay Sensitivity | 14 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 4780 |
| Gene Symbol | NFE2L2 |
| Interassay CV | <12% |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?