missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NrCAM Isoform 3 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
Brand: R&D Systems AF2034
Additional Details : Weight : 0.00970kg
Description
NrCAM Polyclonal specifically detects NrCAM in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
NrCAM | |
Polyclonal | |
Unconjugated | |
Q14CA1 | |
NRCAM | |
Mouse myeloma cell line NS0-derived recombinant human NrCAM isoform 3 Leu30-Asn600 (Ala526Pro) Accession # Q14CA1 | |
100 μg | |
Primary | |
Detects human NrCAM Isoform 3 in direct ELISAs and Western blots. | |
Human | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
bravo, hBravo, KIAA0343Neuronal surface protein Bravo, MGC138845, MGC138846, neuronal cell adhesion molecule, Ng-CAM-related, NgCAM-related cell adhesion molecule, nr-CAM | |
Goat | |
Affinity Purified | |
RUO | |
4897 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only